missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Calcium Chloride, Dihydrate, USP Grade
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 10035-04-8
- Molecular Formula: CaCl2 2H2O
- Molecular Weight: 147.02
Specifications
Specifications
| Chemical Name or Material | Calcium Chloride, Dihydrate, USP Grade |
| CAS | 10035-04-8 |
| Quantity | 1 kg |
| Molecular Formula | CaCl2 - 2H2O |
| Grade | USP |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?