missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™

Product Code. 10306313
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10306313 100 g Poly Bottle 100g
10316313 250 g Poly Bottle 250g
10786634 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10306313 Supplier Fisher BioReagents Supplier No. BP510100
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10035-04-8 | Molecular Formula: Ca Cl2 · 2 H2O

Calcium Chloride Dihydrate (White Crystals to Powder)

Calcium Chloride is suitable for use in the preparation of buffer solutions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8 , 10043-52-4
Molecular Formula Ca Cl2 · 2 H2O

Specifications

Melting Point 175°C
Color Beige-White
pH 4.5 to 6.5
Physical Form Solid
Quantity 100 g
Assay Percent Range 99.0 to 105.0 %
Merck Index 15, 1661
Formula Weight 147.02
Percent Purity 99.0 to 105.0%
Identification Pass Test
Packaging Poly Bottle
Density 0.830g/cm³
Chemical Name or Material Calcium Chloride Dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Calcium Chloride Dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Calcium chloride, dihydrate, Calcium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.