missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Hexahydrate, 98.5%+, Extra Pure, SLR, Fisher Chemical™

Product Code. 12658625 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12658625 5 kg 5kg
12648625 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12658625 Supplier Fisher Chemical Supplier No. C/1250/65
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7774-34-7
Molecular Formula CaCl2H12O6
Molecular Weight (g/mol) 219.07
MDL Number 149616
InChI Key QHFQAJHNDKBRBO-UHFFFAOYSA-L
IUPAC Name calcium hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ca++]

Specifications

Melting Point 30°C
Color Colorless
pH 6.5 to 8.0
Physical Form Solid
Quantity 5 kg
Formula Weight 219.08g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Calcium chloride hexahydrate, 98.5%+, Specified (SLR)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.