Learn More
Calcium chloride hydrate, Puratronic™, 99.9965% (metals basis)
Description
It is used as a food additive, as an electrolyte, it is used as a buffer in pools, an additive in plastics and in fire extinguishers. It is used as a fabric softener, dust control on roads. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Crystalline Aggregates |
| Quantity | 10 g |
| Merck Index | 14,1659 |
| Solubility Information | Soluble in water (5360mg/ml at 20°C),and alcohol. |
| Formula Weight | 110.99 (Anhydrous) |
| Percent Purity | 99.9965% |
| Grade | Puratronic |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 1.71 g/mL |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.