missing translation for 'onlineSavingsMsg'
Learn More

Calcium D-gluconate gel, 2.5% w/w aq. soln.

Product Code. 11425300
Change view
Click to view available options
Quantity:
30 g
60 g
Unit Size:
30g
60g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425300 60 g 60g
11415300 30 g 30g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425300 Supplier Thermo Scientific Alfa Aesar Supplier No. 044542.TJ
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 299-28-5 | C12H22CaO14 | 430.372 g/mol

Calcium D-gluconate gel is used to treat hydrofluoric acid burns. It is used as a cardioprotective agent in hyperkalemia, antidote for black widow spider envenomation and an antidote for magnesium sulfate toxicity. It is also involved in the treatment of hypocalcemia. Further, it is used as skin conditioning as well as provides the source of calcium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-28-5
Molecular Formula C12H22CaO14
Molecular Weight (g/mol) 430.372
MDL Number MFCD00064209
InChI Key NEEHYRZPVYRGPP-IYEMJOQQSA-L
Synonym calcium gluconate, calcium d-gluconate, calciofon, calglucon, glucobiogen, ebucin, calcicol, calcipur, calglucol, dragocal
PubChem CID 9290
IUPAC Name calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]

Specifikationer

Quantity 60 g
Solubility Information Slowly soluble in water. Insoluble in alcohol,acetic acid and organic solvents.
Formula Weight 430.37
Concentration or Composition (by Analyte or Components) 2.5% w/w aq. soln.
Physical Form Gel
Chemical Name or Material Calcium D-gluconate gel

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.