missing translation for 'onlineSavingsMsg'
Learn More

Calcium DL-glycerate dihydrate

Product Code. 11398035
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11398035 1 g 1g
11308045 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11398035

Brand: Thermo Scientific Alfa Aesar L08214.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67525-74-0 | C3H5O4 | 105.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in cold and hot water.

Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67525-74-0
Molecular Formula C3H5O4
Molecular Weight (g/mol) 105.07
MDL Number MFCD00149289,MFCD00149288,MFCD00149289
InChI Key RBNPOMFGQQGHHO-UWTATZPHSA-M
Synonym calciumdl-glycerate
PubChem CID 131883203
IUPAC Name calcium;2,3-dihydroxypropanoic acid;dihydrate
SMILES OC[C@@H](O)C([O-])=O

Specifications

Melting Point ∽140°C (decomposition)
Quantity 1 g
Beilstein 4614104
Merck Index 14,4483
Solubility Information Soluble in cold and hot water.
Formula Weight 286.26 (250.23 Anhydrous)
Chemical Name or Material Calcium DL-glycerate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.