missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, 99%

Product Code. p-7048611
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311638 10 g 10g
11331638 50 g 50g
11321638 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11311638 Supplier Thermo Scientific Alfa Aesar Supplier No. 011022.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-75-5 | CaF2 | 78.08 g/mol

It finds its application in spectroscopy, electronics, lasers, high-temperature dry-film lubricants, emery wheels, certain cements, phosphors, paint pigment, catalyst in wood preservatives, and optical equipment. Calcium fluoride is used as component of electrolyte, fluxing agent for aluminum metallurgy. It is used in glass and fluorescent lamps industry. It has dental applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
PubChem CID 84512
ChEBI CHEBI:35437
IUPAC Name calcium;difluoride
SMILES [F-].[F-].[Ca++]

Specifications

Boiling Point ∼2500°C
Melting Point 1403°C
Physical Form Crystals
Quantity 10 g
Merck Index 14,1667
Solubility Information Soluble in ammonium salts. Insoluble in water.
Formula Weight 78.08
Percent Purity 99%
Odor Odorless
Refractive Index 1.434
Sensitivity Hygroscopic
Density 3.18 g/mL
Chemical Name or Material Calcium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.