missing translation for 'onlineSavingsMsg'
Learn More

Calcium Fluoride, Natural, Honeywell™

Product Code. 15680550 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15680550 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15680550 Supplier Honeywell-Fluka Supplier No. 019055KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Natural,≥95%, powder

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7789-75-5
Physical Form Powder
Quantity 5 kg
Molecular Formula CaF2
MDL Number MFCD00010907
UN Number NONH for all modes of transport
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
SMILES [F-].[F-].[Ca++]
IUPAC Name calcium difluoride
Molecular Weight (g/mol) 78.08
PubChem CID 84512
ChEBI CHEBI:35437
Formula Weight 78.07g/mol
Percent Purity ≥95%
Density 3.18g/mL at 25°C
Chemical Name or Material Calcium Fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.