missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, Puriss., meets analytical spec. of DAB, 99.0-102.0%, Honeywell Fluka™

Product Code. p-7071691 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
1 kg
40 kg
Unit Size:
100g
1kg
40kg
Product Code. Quantity unitSize
15620540 100 g 100g
15630540 1 kg 1kg
15640540 40 kg 40kg
3 options
This item is not returnable. View return policy

Product Code. 15620540

Brand: Honeywell-Fluka 01123100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., meets analytical specification of DAB, 99.0-102.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
PubChem CID 84512
ChEBI CHEBI:35437
IUPAC Name calcium;difluoride
SMILES [F-].[F-].[Ca++]

Specifications

Quantity 100 g
UN Number NONH for all modes of transport
Formula Weight 78.07g/mol
Percent Purity 99 to 102%
Grade puriss.
Density 3.18g/mL at 25°C
Chemical Name or Material Calcium Fluoride, Meets analytical specification of DAB
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.