missing translation for 'onlineSavingsMsg'
Learn More

Calcium formate, 98%

Product Code. 11413168
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413168 500 g 500g
11403168 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413168 Supplier Thermo Scientific Alfa Aesar Supplier No. A18916.36
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 544-17-2 | C2H2CaO4 | 130.112 g/mol

An industrial chemical compound and faster setting, early strength gain accelerating admixture for concrete or mortars. It is used in the building industry, in the production of feedstuff mixtures, for silage and in the tannery. Also used in leather tanning, producing formic acid, concrete cure accelerator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An industrial chemical compound and faster setting, early strength gain accelerating admixture for concrete or mortars. It is used in the building industry, in the production of feedstuff mixtures, for silage and in the tannery. Also used in leather tanning, producing formic acid, concrete cure accelerator.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-17-2
Molecular Formula C2H2CaO4
Molecular Weight (g/mol) 130.112
MDL Number MFCD00036108
InChI Key CBOCVOKPQGJKKJ-UHFFFAOYSA-L
Synonym calcium formate, calcium diformate, formic acid, calcium salt, calcoform, unii-np3jd65npy, mravencan vapenaty czech, calcium formate ca hco2 2, formic acid calcium salt, np3jd65npy, formic acid, calcium salt 2:1
PubChem CID 10997
ChEBI CHEBI:81851
IUPAC Name calcium;diformate
SMILES C(=O)[O-].C(=O)[O-].[Ca+2]

Specifications

Melting Point >300°C
Density 2.02
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 130.12
Percent Purity 98%
Chemical Name or Material Calcium formate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.