missing translation for 'onlineSavingsMsg'
Learn More

Calcium Gluconate, TRC

Product Code. 30133394
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133394 50 g 50g
17978094 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133394 Supplier Toronto Research Chemicals Supplier No. TRCC14550050G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-28-5
Molecular Formula 2 C6 H11 O7 . Ca
Molecular Weight (g/mol) 430.37
Synonym Calcium gluconate anhydrous, Anhydrous calcium d-gluconate, Calcium digluconate, D-Gluconic acid, calcium salt (2:1), Gluconic acid, calcium salt (2:1), D- (8CI), Biocal, Calcicol, Calciofon, Calcipur, Calcium D-gluconate, Calcium gluconate, Calcium hexagluconate, Calglucol, Calglucon, Dragocal, Ebucin, Glucal, Glucobiogen, Kalpren, Novocal
IUPAC Name calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[Ca]OC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

Specifications

InChI Formula InChI=1S/2C6H12O7.Ca/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;/h2*2-5,7-11H,1H2,(H,12,13);/q;;+2/p-2/t2*2-,3-,4+,5-;/m11./s1
Formula Weight 430.0635
Percent Purity >95%
Quantity 50 g
Chemical Name or Material Calcium gluconate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.