missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydride, Technical, Fisher Chemical

Product Code. 11408952 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408952 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408952 Supplier Fisher Chemical Supplier No. C/1620/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 675°C
CAS 7789-78-8
Color Gray
Physical Form Solid
Quantity 100 g
Molecular Formula CaH2
MDL Number 10897
InChI Key FAQLAUHZSGTTLN-UHFFFAOYSA-N
SMILES [CaH2]
IUPAC Name calcium dihydride
Molecular Weight (g/mol) 42.09
Formula Weight 42.09g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.