missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydroxide, 95+%, ACS reagent

Product Code. 10721632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10721632 1 kg Plastic bottle 1kg
10249243 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10721632 Supplier Thermo Scientific Acros Supplier No. 403850010
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
IUPAC Name calcium dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Boiling Point 2850.0°C
Melting Point 580.0°C
Color White
Physical Form Amorphous Powder
Quantity 1 kg
Assay Percent Range 95+%
Linear Formula Ca(OH)2
Merck Index 15, 1675
Solubility Information Solubility in water: 1.65g/L (20°C). Other solubilities: soluble in acids (evolution of heat) and nh4cl sol, soluble in glycerol and sugar, insoluble in alcohol
Formula Weight 74.09
Percent Purity ≥95%
Grade ACS Reagent
Packaging Plastic bottle
Density 2.24g/mL
Chemical Name or Material Calcium hydroxide, For analysis ACS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Calcium hydroxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.