missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydroxide, 95%

Product Code. 11460503
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460503 250 g 250g
11450503 1000 g 1000g
11470503 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460503 Supplier Thermo Scientific Alfa Aesar Supplier No. A12650.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Calcium hydroxide is used as a flocculant in water and sewage treatment. It is also used in fresh water treatment to raise the pH of the water thereby controlling the corrosion of pipes. It is widely used in the production of calcium stearate. It is an active ingredient in whitewash, mortar and plaster. This compound is also used in the petroleum refining industry for the preparation of additives to oils and in the preparation of dry mixes for painting and decorating. It finds application as an antifungal and antimicrobial preservative for vegetables in storage.

Solubility
Soluble in glycerol and acids. Slightly soluble in water.Insoluble in alcohol.

Notes
Air sensitive. Incompatible with strong acids.Incompatible with maleic anhydride, phosphorous, nitroethane, nitromethane, nitorparaffins, nitropropane and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
IUPAC Name calcium;dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Quantity 250 g
Linear Formula Ca(OH)2
UN Number UN3262
Merck Index 14,1673
Solubility Information Soluble in glycerol and acids. Slightly soluble in water.Insoluble in alcohol.
Formula Weight 74.09
Percent Purity 95%
Odor Odorless
Sensitivity Air Sensitive
Density 2.24
Chemical Name or Material Calcium hydroxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Calcium hydroxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: calcium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.