missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydroxide, 99.995%, (trace metal basis)

Product Code. 11914161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11914161 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11914161 Supplier Thermo Scientific Acros Supplier No. 385610050
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 2850.0°C
Melting Point 580.0°C
CAS 1305-62-0
Color White
Physical Form Crystals or Powder
Quantity 5 g
Assay Percent Range 54% min. (Ca) (Complexometry)
Molecular Formula CaH2O2
Linear Formula Ca(OH)2
MDL Number MFCD00010901
Merck Index 15,1675
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
Solubility Information Solubility in water: 1.65g/L (20°C). Other solubilities: soluble in acids (evolution of heat) and nh4cl sol, soluble in glycerol and sugar, insoluble in alcohol
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES [OH-].[OH-].[Ca++]
IUPAC Name calcium;dihydroxide
Molecular Weight (g/mol) 74.09
PubChem CID 6093208
ChEBI CHEBI:31341
Formula Weight 74.09
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Density 2.24g/mL
Chemical Name or Material Calcium hydroxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Calcium hydroxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.