missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydroxide, Puriss., meets analytical specification of FCC, Ph. Eur., USP, 95-100.5% (complexometric), Solstice

Product Code. 15661090 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Paper / Poly Bag
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15661090 25 kg Paper / Poly Bag 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15661090 Supplier Honeywell Fluka Supplier No. 1203825KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1305-62-0
CAS Max % 100.0000%
Quantity 25 kg
Assay Percent Range 95 to 100.5%
Molecular Formula CaH2O2
Linear Formula Ca(OH)2
MDL Number MFCD00010901
UN Number NONH for all modes of transport
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES [OH-].[OH-].[Ca++]
IUPAC Name calcium;dihydroxide
Molecular Weight (g/mol) 74.09
PubChem CID 6093208
ChEBI CHEBI:31341
Formula Weight 74.09g/mol
Percent Purity 95 to 100.5% (complexometric)
Grade puriss.
Packaging Paper / Poly Bag
Density 2.24g/mL at 25°C
Chemical Name or Material Calcium hydroxide, Puriss., meets analytical specification of FCC, Ph. Eur., USP, 95-100.5% (complexometric)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.