missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, Reag. Ph. Eur., puriss. p.a., Solstice

Product Code. 15653300
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
6 x 1 kg
Unit Size:
100g
1kg
25kg
500g
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15653300 6 x 1 kg Pack of 6
15613300 100 g 100g
15643300 500 g 500g
15623300 1 kg 1kg
15633300 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15653300 Supplier Honeywell-Fluka Supplier No. 312196X1KG
  • ONLINE EXCLUSIVE PRICE
    603.00 €
    545.13 € / Pack of 6
    Save 57.87 €
    9% Off
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., Reag. Ph. Eur.,≥96%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
IUPAC Name calcium;dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Quantity 6 x 1 kg
Linear Formula Ca(OH)2
UN Number NONH for all modes of transport
Formula Weight 74.09
Percent Purity ≥96%
Grade puriss. p.a.
Density 2.24g/mL at 25°C
Chemical Name or Material Calcium Hydroxide, Reag. Ph. Eur.
compliance-icons compliance-icons
Product Identifier
  • Calcium hydroxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Calcium dihydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.