missing translation for 'onlineSavingsMsg'
Learn More

Calcium hypochlorite, tech., granular, 65% available chlorine

Product Code. 10626291 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10626291 1 kg Plastic bottle 1kg
10195310 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10626291 Supplier Thermo Scientific Acros Supplier No. 300340010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7778-54-3 | CaCl2O2 | 142.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-54-3
Molecular Formula CaCl2O2
Molecular Weight (g/mol) 142.98
MDL Number MFCD00010900
InChI Key ZKQDCIXGCQPQNV-UHFFFAOYSA-N
Synonym calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor
PubChem CID 24504
IUPAC Name calcium;dihypochlorite
SMILES [O-]Cl.[O-]Cl.[Ca+2]

Specifications

Melting Point 100°C
Color Green or White
pH ∼11.6 (1% aq. soln.)
Physical Form Granular Solid or Granules
Quantity 1 kg
Packaging Plastic bottle
Linear Formula Ca(OCl)2
Fieser 11,107
Merck Index 15,1676
Solubility Information Solubility in water: 200g/L (20°C) (decomposes). Other solubilities: decomposes in alcohol
Formula Weight 142.98
Percent Purity ≥65% (available Cl)
Grade Technical
Chemical Name or Material Calcium hypochlorite, tech.
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Calcium hypochlorite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • H272-May intensify fire; oxidiser.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.