missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hypochlorite (Technical Grade), TRC

Product Code. 30336892
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30336892

Brand: Toronto Research Chemicals TRCC1455405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium Hypochlorite (Technical Grade)
CAS 7778-54-3
Molecular Formula Ca(OCl)2
Quantity 5 g
Synonym Hypochlorous Acid Calcium Salt, B-K Powder, Bleaching Powder, Ca(ClO)2, Calcium Hypochlorite, Calcium Oxychloride, Caporit, Chemichlon G, Chloride of Lime, Chlorinated Lime, Chloro Lime Chemical, Eusol BPCHTH (bleaching agent), HTH VAN, Lime Chloride, Losantin, Nisso Hi-Chlon, Nisso Hi-chlon LC, Perfect PA, Solvox KS, Super Zappit, T-Eusol, Zappit, Calcium Di(oxychloride)
InChI Formula InChI=1S/Ca.2ClO/c;2*1-2/q+2;2*-1
SMILES [Ca+2].[O-]Cl.[O-]Cl
IUPAC Name calcium;dihypochlorite
Molecular Weight (g/mol) 142.98
Formula Weight 141.8901
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.