missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Hypochlorite (Technical Grade), TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Calcium Hypochlorite (Technical Grade) |
| CAS | 7778-54-3 |
| Molecular Formula | Ca(OCl)2 |
| Quantity | 50 g |
| Synonym | Hypochlorous Acid Calcium Salt, B-K Powder, Bleaching Powder, Ca(ClO)2, Calcium Hypochlorite, Calcium Oxychloride, Caporit, Chemichlon G, Chloride of Lime, Chlorinated Lime, Chloro Lime Chemical, Eusol BPCHTH (bleaching agent), HTH VAN, Lime Chloride, Losantin, Nisso Hi-Chlon, Nisso Hi-chlon LC, Perfect PA, Solvox KS, Super Zappit, T-Eusol, Zappit, Calcium Di(oxychloride) |
| InChI Formula | InChI=1S/Ca.2ClO/c;2*1-2/q+2;2*-1 |
| SMILES | [Ca+2].[O-]Cl.[O-]Cl |
| IUPAC Name | calcium;dihypochlorite |
| Molecular Weight (g/mol) | 142.98 |
| Formula Weight | 141.8901 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction