missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodide, ultra dry, 99.995% (metals basis)

Product Code. 15447395
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447395 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15447395 Supplier Thermo Scientific Alfa Aesar Supplier No. 047121.14
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10102-68-8 | CaI2 | 293.887 g/mol

It is used in photography. It is also used in cat food as a source of iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-68-8
Molecular Formula CaI2
Molecular Weight (g/mol) 293.887
MDL Number MFCD00010910
InChI Key UNMYWSMUMWPJLR-UHFFFAOYSA-L
Synonym calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
PubChem CID 5462722
ChEBI CHEBI:81718
IUPAC Name calcium;diiodide
SMILES [Ca+2].[I-].[I-]

Specifications

Boiling Point 1100°C
Melting Point 784°C
Color White to Yellow
Physical Form Beads
Quantity 25 g
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Density 3.956 g/mL
Merck Index 14,1677
Solubility Information Soluble in water (64.6g/100 mL (0°C)). Soluble in acetone and alcohols.
Formula Weight 293.89
Percent Purity 99.995%
Assay (metals basis)
Chemical Name or Material Calcium iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.