missing translation for 'onlineSavingsMsg'
Learn More

Calcium L-lactate pentahydrate, 98%

Product Code. 10666461 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10666461 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10666461 Supplier Thermo Scientific Acros Supplier No. 250200010
Chemical Monthly Promotion
Use promo code 28093
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 865717-20-0 | C6H10CaO6·5H2O | 308.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 240.0°C
CAS 865717-20-0, 28305-25-1
Color White
pH 6.0 to 8.0 (5% aq. soln.)
Physical Form Powder
Quantity 1 kg
Assay Percent Range 97.5% min. (on dry substance) (Complexometry)
Molecular Formula C6H10CaO6·5H2O
MDL Number MFCD00287281
Beilstein 03, IV, 636
Merck Index 14, 1678
Synonym Lactic acid, calcium salt pentahydrate, (S)-2-Hydroxypropanoic acid, calcium salt pentahydrate
Solubility Information Solubility in water: 90g/L (25°C). Other solubilities: practically insoluble in alcohol
InChI Key YNIXORYWSZWLSA-VFGZHDCSSA-N
SMILES CC(C(=O)O)O.CC(C(=O)O)O.O.O.O.O.O.[Ca]
IUPAC Name calcium;(2S)-2-hydroxypropanoic acid;pentahydrate
Molecular Weight (g/mol) 308.29
PubChem CID 131886284
Formula Weight 308.29
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 22.0% to 27.0%
Packaging Plastic bottle
Chemical Name or Material Calcium L-lactate pentahydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.