missing translation for 'onlineSavingsMsg'
Learn More

Calcium metaborate, 99.992% (metals basis)

Product Code. 11342088
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342088 100 g 100g
11322088 5 g 5g
11332088 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342088 Supplier Thermo Scientific Alfa Aesar Supplier No. 012988.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13701-64-9 | B2CaO4 | 125.694 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 13701-64-9
Molecular Formula B2CaO4
Molecular Weight (g/mol) 125.694
MDL Number MFCD00049397
InChI Key ILOKQJWLMPPMQU-UHFFFAOYSA-N
Synonym calcium metaborate, 2bo2.ca, calcium dioxoborinate, dimetaboric acid calcium salt, calcium 2+ diboron oxide hydroxide
PubChem CID 10290800
IUPAC Name calcium;oxido(oxo)borane
SMILES B(=O)[O-].B(=O)[O-].[Ca+2]

Spezifikation

Color White
Physical Form Powder
Quantity 100 g
Linear Formula Ca(BO2)2
Formula Weight 125.7
Percent Purity 99.992%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Calcium metaborate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.