missing translation for 'onlineSavingsMsg'
Learn More

Calcium molybdenum oxide, Puratronic™, 99.9965% (metals basis), Thermo Scientific™

Product Code. 11310929
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11310929 5 g 5g
11300929 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310929 Supplier Thermo Scientific Alfa Aesar Supplier No. 010682.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-82-4
Molecular Formula CaMoO4
Molecular Weight (g/mol) 200.02
MDL Number MFCD00015976
InChI Key BIOOACNPATUQFW-UHFFFAOYSA-N
Synonym calcium molybdate, molybdate, calcium, calcium molybdate vi, unii-q03c83v2an, calcium molybdenum oxide, molybdic acid h2moo4 , calcium salt 1:1, molybdate moo42-, calcium 1:1 , t-4, molybdate moo42-, calcium 1:1 , beta-4, calcium dioxido dioxo molybdenum, calcium 2+ molybdate
PubChem CID 24620
IUPAC Name calcium;dioxido(dioxo)molybdenum
SMILES [Ca++].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 965°C
Physical Form Powder
Quantity 5 g
Merck Index 14,1681
Formula Weight 200.01
Percent Purity 99.9965%
Grade Puratronic
Odor Odorless
Density 4.38 g/mL to 4.53 g/mL
Assay (metals basis)
Chemical Name or Material Calcium molybdenum oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.