missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate ACS AR Crystal, Macron Fine Chemicals™

Product Code. 15538694
Change view
Click to view available options
Quantity:
500 g
Packaging:
POLYSTORMOR
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15538694 500 g POLYSTORMOR 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15538694 Supplier Avantor Macron Supplier No. 423604
  • ONLINE EXCLUSIVE PRICE
    146.00 €
    127.08 € / 500g
    Save 18.92 €
    12% Off
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Physical Form Crystalline
Quantity 500 g
Formula Weight 236.15
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Calcium Nitrate, 4-Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.