missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate Tetrahydrate, ≥99%, (ACS Reagent), MP Biomedicals

Product Code. 11402401 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11402401 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11402401 Supplier MP Biomedicals Supplier No. 0219380180
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS grade reagent for industrial applications and culture media, commonly used to study bacterial growth and cell fixation.

Specifications

CAS 13477-34-4
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range ≥99%
Molecular Formula CaH8N2O10
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
SMILES O.[Ca++].[O-][N+]([O-])=O
IUPAC Name calcium bis(nitric acid) tetrahydrate
Molecular Weight (g/mol) 236.15
PubChem CID 16211656
ChEBI CHEBI:86159
Formula Weight 236.2
Grade ACS Reagent
Chemical Name or Material Calcium Nitrate Tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.