missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitrate tetrahydrate, Puriss., ≥98%, Honeywell

Product Code. 15671090 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 kg
Packaging:
Paper / Poly Bag
Unit Size:
25kg
This item is not returnable. View return policy

Product Code. 15671090

Brand: Honeywell Research Chemicals 1204025KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 13477-34-4
CAS Max % 100.0000%
Quantity 25 kg
Molecular Formula CaH8N2O10
Linear Formula Ca(NO3)2 · 4H2O
MDL Number MFCD00149604
UN Number NONH for all modes of transport
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
SMILES O.[Ca++].[O-][N+]([O-])=O
IUPAC Name calcium bis(nitric acid) tetrahydrate
Molecular Weight (g/mol) 236.15
PubChem CID 16211656
ChEBI CHEBI:86159
Formula Weight 236.15g/mol
Percent Purity >98%
Grade puriss.
Packaging Paper / Poly Bag
Chemical Name or Material Calcium nitrate tetrahydrate, Puriss., ≥98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.