missing translation for 'onlineSavingsMsg'
Learn More

Calcium Oxide, Solstice
BIOPHARMA

Product Code. 15691090 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
20 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
20kg
5kg
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15691090 2.5 kg 2.5kg
15681090 1 kg 1kg
15611100 5 kg 5kg
15621100 6 x 1 kg Pack of 6
15601100 20 kg 20kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15691090 Supplier Honeywell-Fluka Supplier No. 120472.5KG
  • ONLINE EXCLUSIVE PRICE
    70.00 €
    63.15 € / 2.5kg
    Save 6.85 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of FCC, 96-100.5% (ex ignited substance), powder (fine)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
SMILES [O--].[Ca++]

Specifications

Physical Form Fine Powder
Quantity 2.5 kg
Density 3.3g/mL at 25°C
Linear Formula CaO
UN Number 1910
IUPAC Name oxocalcium
Formula Weight 56.08g/mol
Percent Purity 96 to 100.5% (ex ignited substance)
Grade puriss.
Chemical Name or Material Calcium Oxide, Meets analytical specification of FCC
compliance-icons compliance-icons
Product Identifier
  • Calcium oxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Calcium oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.