missing translation for 'onlineSavingsMsg'
Learn More

Calcium peroxide, typically 65%

Product Code. 11358007
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358007 500 g 500g
11348007 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358007 Supplier Thermo Scientific Alfa Aesar Supplier No. 021157.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rubber stabilizer | CAS: 1305-79-9 | CaO2 | 72.076 g/mol

Calcium peroxide is used as a rubber stabilizer, used in different areas of industry and agriculture. In agriculture it is used as an oxygen fertilizer, and is also used in the presowing treatments of rice seed. It is used as flour bleaching agent and improving agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-79-9
Molecular Formula CaO2
Molecular Weight (g/mol) 72.076
MDL Number MFCD00015988
InChI Key LHJQIRIGXXHNLA-UHFFFAOYSA-N
Synonym calcium peroxide, calcium dioxide, calper, calper g, unii-7fro2eno91, peroxidocalcium, calciumperoxid, kalziumperoxid, calcium peroxide ca o2, hsdb 965
PubChem CID 14779
ChEBI CHEBI:48233
IUPAC Name calcium;peroxide
SMILES [O-][O-].[Ca+2]

Specifications

Physical Form Powder
Quantity 500 g
UN Number UN1457
Merck Index 14,1689
Solubility Information Slightly soluble in water; Soluble in acids
Formula Weight 72.08
Percent Purity 65% (typically)
Odor Odorless
Sensitivity Moisture sensitive
Chemical Name or Material Calcium peroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.