missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate, 96%, pure, mixture of Calcium phosphates

Product Code. 12065390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12065390 1 kg 1kg
12075390 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12065390 Supplier Thermo Scientific Acros Supplier No. 446390010
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7758-87-4 | Ca3O8P2 | 310.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-87-4
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.18
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
Synonym calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic
PubChem CID 24456
ChEBI CHEBI:9679
IUPAC Name tricalcium;diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]

Specifications

Melting Point 1670.0°C
Quantity 1 kg
Packaging Glass bottle
Assay Percent Range 95% min. (Complexometry)
Linear Formula Ca3(PO4)2
Merck Index 15,1696
Solubility Information Solubility in water: 0.1g/L (25°C)
Formula Weight 310.18
Percent Purity 96%
Grade Pure
Chemical Name or Material Calcium phosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.