missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate, for analysis, 35-40% (Ca), mixture of Calcium phosphates

Product Code. 12604967 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12604967 1 kg 1kg
12614967 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12604967 Supplier Thermo Scientific Acros Supplier No. 213480010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7758-87-4 | Ca3O8P2 | 310.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-87-4
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.18
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
Synonym calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic
PubChem CID 24456
ChEBI CHEBI:9679
IUPAC Name tricalcium;diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]

Specifications

Melting Point 1670.0°C
Quantity 1 kg
Assay Percent Range 35-40%
Linear Formula Ca3(PO4)2
Merck Index 15,1696
Solubility Information Solubility in water: 0.1g/L (25°C)
Formula Weight 310.18
Percent Purity 35 to 40% (Ca)
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Calcium phosphate, c.p.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.