missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate Dibasic Dihydrate, Honeywell™

Product Code. 15660720 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15660720 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15660720 Supplier Honeywell-Fluka Supplier No. 042311KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7789-77-7
Quantity 1 kg
Molecular Formula CaH5O6P
Linear Formula CaHPO4 · 2H2O
MDL Number MFCD00149621
UN Number NONH for all modes of transport
Synonym calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen
InChI Key XAAHAAMILDNBPS-UHFFFAOYSA-L
SMILES O.O.OP(=O)([O-])[O-].[Ca+2]
IUPAC Name calcium;hydrogen phosphate;dihydrate
Molecular Weight (g/mol) 172.086
PubChem CID 104805
ChEBI CHEBI:4496
Formula Weight 172.09g/mol
Percent Purity 98 to 102.5%
Grade puriss.
Chemical Name or Material Calcium Phosphate Dibasic Dihydrate, Meets analytical specification of Ph. Eur.BP, USP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.