missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Phosphate Dibasic, puriss. p.a., >98.5% (KT), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
Unit Size:
250g
50g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Tekniske data
| Melting Point | ≥450°C |
| Color | White |
| Physical Form | Anhydrous |
| Quantity | 50 g |
| Linear Formula | CaHPO4 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 136.06g/mol |
| Percent Purity | >98.5% (KT) |
| Grade | puriss. p.a. |
| Packaging | Poly Bottle |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion