missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate tribasic, powder NF, FCC, Macron Fine Chemicals™

Product Code. 15508284
Change view
Click to view available options
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15508284 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15508284 Supplier Macron Fine Chemicals Supplier No. 428003
  • ONLINE EXCLUSIVE PRICE
    340.80 €
    304.23 € / 500g
    Save 36.57 €
    10% Off
 Restricted Item
Estimated Shipment: 19-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Meets F.C.C. Requirements
  • GMP Manufactured Product
  • Meets N.F. Requirements
  • Food GMP Manufactured Product
  • CAUTION: For Manufacturing, processing or repackaging
  • Bulk Pharmaceutical Chemical
  • Bulk Food Chemical
  • No Class 1,2,3 or other solvents are used or produced in themanufacturing or purification of the product.
  • Metallic Residues: No metal catalysts or metal reagents, as defined by EMA Guideline EMEA/CHMP/SWP/4446/2000, are used in the production of
    this material.

Specifications

CAS 1306-06-5
Quantity 500 g
Assay Percent Range 34.0 to 40.0%
Molecular Formula Ca5HO13P3
MDL Number MFCD00010904
InChI Key XYJRXVWERLGGKC-UHFFFAOYSA-D
SMILES [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
IUPAC Name pentacalcium hydroxide triphosphate
Molecular Weight (g/mol) 502.31
Identification Passes Test
Loss on Ignition ≤ 8.0%
Packaging Mall POLYSTORMOR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.