missing translation for 'onlineSavingsMsg'
Learn More

Calcium Pyruvate (Technical Grade), TRC

Product Code. 30344323
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30344323 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344323 Supplier Toronto Research Chemicals Supplier No. TRCC1456582.5G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium Pyruvate (Technical Grade)
CAS 52009-14-0
Molecular Formula C6H6O6Ca
Quantity 2.5 g
Synonym Propanoic acid, 2-oxo-, calcium salt (2:1), Propanoic acid, 2-oxo-, calcium salt (9CI), Calcium pyruvate, Calcium 2-oxopropanoate, Calcium α-ketopropionate, Pyruvic acid calcium salt, 2-Oxopropanoic acid hemicalcium salt
InChI Formula InChI=1S/2C3H4O3.Ca/c2*1-2(4)3(5)6;/h2*1H3,(H,5,6);/q;;+2/p-2
SMILES [Ca+2].CC(=O)C(=O)[O-].CC(=O)C(=O)[O-]
IUPAC Name calcium;2-oxopropanoate
Molecular Weight (g/mol) 214.18
Formula Weight 213.979
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.