missing translation for 'onlineSavingsMsg'
Learn More

Calcium silicate, meta, 99% (metals basis), Thermo Scientific™

Product Code. 15161995
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15161995 100 g 100g
11308747 25 g 25g
11398737 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15161995 Supplier Thermo Scientific Alfa Aesar Supplier No. 035843.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-39-0
Molecular Formula CaO3Si
Molecular Weight (g/mol) 116.16
MDL Number MFCD00015979
InChI Key OYACROKNLOSFPA-UHFFFAOYSA-N
Synonym calcium metasilicate, wollastonite, aedelforsite, casiflux, cab-o-lite, gillebachite, wollastokup, calcium silicon oxide, florite, fuwalip
PubChem CID 26370
IUPAC Name calcium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[Ca+2]

Specifications

Melting Point 1540°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%
Solubility Information Insoluble in water.
Formula Weight 116.16
Odor Odorless
Appearance White powder
Density 2.9 g/cm3 at 20°C
Chemical Name or Material Calcium silicate, meta
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.