missing translation for 'onlineSavingsMsg'
Learn More

Calcium Stearate, Technical, Fisher Chemical

Product Code. 10540911 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10540911 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10540911 Supplier Fisher Chemical Supplier No. C/2240/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 145°C
CAS 1592-23-0
Color White
Physical Form Solid
Quantity 500 g
Molecular Formula C36H70CaO4
MDL Number 36390
InChI Key CJZGTCYPCWQAJB-UHFFFAOYSA-L
SMILES [Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
IUPAC Name calcium dioctadecanoate
Molecular Weight (g/mol) 607.03
Formula Weight 607.02g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.