missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate, anhydrous, Puratronic™, 99.993% (metals basis)

Product Code. 11326138
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326138 2 g 2g
11336138 10 g 10g
11346138 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326138 Supplier Thermo Scientific Alfa Aesar Supplier No. 011116.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

Calcium sulfate, anhydrous is used as adhesives and sealant chemical, Fillers, Fuels and fuel additives, Intermediates, Process regulators, Agricultural Products, Building/Construction Materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
Synonym calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium;sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1450°C
Color White
Physical Form Powder
Quantity 2 g
Linear Formula CaSO4
Merck Index 14,1706
Solubility Information Soluble in water,ammonium salts,sodium thiosulfate & glycerin.
Formula Weight 136.14
Percent Purity 99.993%
Grade Puratronic
Odor Odorless
Sensitivity Hygroscopic
Density 2.960 g/mL
Assay (metals basis)
Chemical Name or Material Calcium sulfate, Anhydrous
Show More Show Less

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.