missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate Dihydrate, 98%, ACS Reagent, Honeywell Fluka™

Product Code. 15642750 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
500g
Product Code. Quantity Packaging unitSize
15642750 100 g Poly Bottle 100g
15662750 500 g Poly Bottle 500g
15652750 1 kg Poly Bottle 1kg
3 options
This item is not returnable. View return policy

Product Code. 15642750

Brand: Honeywell-Fluka 255548100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Solid
Quantity 100 g
Linear Formula CaSO4 · 2H2O
UN Number NONH for all modes of transport
IUPAC Name calcium;sulfate;dihydrate
Formula Weight 172.17g/mol
Percent Purity 98%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Density 2.320g/cm3
Chemical Name or Material Calcium Sulfate Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.