missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate dihydrate, 98+%, for analysis

Product Code. 10462031 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10462031 25 g Plastic bottle 25g
10398690 500 g Plastic bottle 500g
11348611 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10462031 Supplier Thermo Scientific Acros Supplier No. 225270250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10101-41-4 | CaH4O6S | 172.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium;sulfate;dihydrate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 98.0 to 102% (Complexometry)
Linear Formula CaSO4·2H2O
Fieser 01,107
Merck Index 14,1706
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents
Formula Weight 172.17
Percent Purity ≥98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Calcium sulfate dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.