missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate Dihydrate, ≥99%, ReagentPlus™ Grade, Honeywell™

Product Code. 15647750 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15647750 500 g Poly bottle 500g
15637750 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15647750 Supplier Honeywell Research Chemicals Supplier No. C3771500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Color Colorless
Physical Form Solid
Quantity 500 g
Linear Formula CaSO4 · 2H2O
UN Number NONH for all modes of transport
IUPAC Name calcium dihydrate sulfate
Formula Weight 172.17g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Calcium sulfate dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.