missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate Dihydrate, ≥99%, ReagentPlus™ Grade, Honeywell™

Product Code. p-7120289 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
500g
Product Code. Quantity Packaging unitSize
15647750 500 g Poly bottle 500g
15637750 1 kg Poly bottle 1kg
2 options
This item is not returnable. View return policy

Product Code. 15647750

Brand: Honeywell Research Chemicals C3771500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Color Colorless
Physical Form Solid
Quantity 500 g
Linear Formula CaSO4 · 2H2O
UN Number NONH for all modes of transport
IUPAC Name calcium dihydrate sulfate
Formula Weight 172.17g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Calcium sulfate dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.