missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate dihydrate, Puriss., meets analytical spec. of NF, E 516, 99.0-101.0% (based on anhydrous substance), Solstice
BIOPHARMA

Product Code. 15651100
Change view
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
Unit Size:
1kg
25kg
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15651100 6 x 1 kg Pack of 6
15631100 1 kg 1kg
15641100 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15651100 Supplier Honeywell-Fluka Supplier No. 120566X1KG
  • ONLINE EXCLUSIVE PRICE
    293.00 €
    264.17 € / Pack of 6
    Save 28.83 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium;sulfate;dihydrate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

CAS Max % ≤100.0000%
Quantity 6 x 1 kg
Assay Percent Range 99.0 to 101.0%
Linear Formula CaSO4 · 2H2O
Grade puriss.
Packaging Poly Bottle
Density 2.320 g/cm3
Chemical Name or Material Calcium sulfate dihydrate, Puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.