missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfide, 99% (metals basis excluding Mg), Mg <1%

Product Code. 11335867
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
missing translation for 'unitSize'
100g
25g
This item is not returnable. View return policy

Product Code. 11335867

missing translation for 'mfr': Thermo Scientific Alfa Aesar 043646.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20548-54-3 | CaS | 72.14 g/mol

Calcium sulfide is used in phosphors, lubricant additives and luminous paints. It reacts with elemental sulfur to give a lime-sulfur. Further, it is used in ore dressing and flotation agent. In addition to this, it is used as a depilatory in the tanning industry and in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20548-54-3
Molecular Formula CaS
Molecular Weight (g/mol) 72.14
MDL Number MFCD00015990
InChI Key AGVJBLHVMNHENQ-UHFFFAOYSA-N
PubChem CID 30182
IUPAC Name calcium sulfanediide
SMILES [S--].[Ca++]

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 25 g
UN Number UN3134
Merck Index 14,1707
Solubility Information Slightly soluble in water. Insoluble in alcohol.
Formula Weight 72.14
Percent Purity 99%
Odor Stench
Sensitivity Moisture sensitive
Density 2.5 g/mL
Assay (metals basis excluding Mg), Mg <1%
Chemical Name or Material Calcium sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.