Learn More
BINDER™ Calibration Certificate, Temperature
Calibration certificate, temperature - for temperature, measurement in center of chamber at specified temperature
Brand: BINDER™ 8012-1567
Additional Details : Weight : 0.00200kg
Description
- Measurement at specified temperature
- Including certificate
- Various points of measurement
Specifications
BINDER Chambers (Models MKFT 115, MKFT 240, MKFT 720) | |
Calibration certificate, temperature - temperature measurement incl. certificate, 9 measuring points (at specified temperature) | |
Calibration Certificate (Temperature) | |
Temperature Measurement |