missing translation for 'onlineSavingsMsg'
Learn More

Draeger Calibration Gas, Sulfur Dioxide in Nitrogen

Product Code. 15987785
Change view
Click to view available options
Volume (Metric) Gas:
58 L
Unit Size:
60 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Gas unitSize
15987785 58 L 60 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15987785 Supplier Draeger Supplier No. 6810645

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Single gas, sulfur dioxide SO2 in nitrogen 10ppm supplied in a non-refillable cylinder.

  • Non-refillable cylinder
  • Cylinders are small, light and recyclable, disposable
  • Sulfur dioxide SO2 in 10ppm nitrogen
  • Requires regulator and delivery tubing if not used with a Draeger bump test station
  • Single gas mixtures of reliable, constant quality

Specifications

Gas Type Sulfur Dioxide in 10 ppm Nitrogen
Volume (Metric) Gas 58 L
For Use With (Application) Calibration and Bump Testing, Portable Gas Detection
Includes Requires a regulator and delivery tubing if not used with a Draeger bump test station
compliance-icons
Product Identifier
  • 60LT Sulfur Dioxide SO2/nitrogen 10ppm calibration
Hazard Category
  • Gases under pressure
Hazard Statement
  • H280-Contains gas under pressure; may explode if heated.
Precautionary Statement
  • P103-Read label before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P233-Keep container tightly closed.
  • P376-Stop leak if safe to do so.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P410+P403-Protect from sunlight Store in a well-ventilated place.
  • P412-Do not expose to temperatures exceeding 50°C/122°F.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sulphur dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.