Learn More
Applied Biosystems™ Calibration Kit, 384-well
Product Code. 15840393 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 384 well plate
Unit Size:
Each
Product Code. Quantity Unit Size
15840393 1 x 384 well plate Each
1 options
Product Code. 15840393 Supplier Applied Biosystems™ Supplier No. 4432476
  • ONLINE EXCLUSIVE PRICE
    3415.00 €
    2049.00 € / Pack of 1
    Save 1366.00 €
    40% Off
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The 384-Well Calibration Kit is used to maintain your real-time PCR instrument with 384-well block
  • The 384-Well Calibration Kit is used to maintain your real-time PCR instrument with 384-well block.
  • The kit includes region of interest (ROI), background, and dye calibrations, as well as RNaseP verification.
  • Please see your instrument user guide for full instructions on use.
TRUSTED_SUSTAINABILITY
Specifications
Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Product Type Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 x 384 well plate
Calibration Type Spectral Calibration
PCR Method qPCR