missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Calibration Kit, 96-well

Product Code. 15860393 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 96 well plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860393 1 x 96 well plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15860393 Supplier Applied Biosystems™ Supplier No. 4432494
  • ONLINE EXCLUSIVE PRICE
    3455.00 €
    2073.00 € / Pack of 1
    Save 1382.00 €
    40% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The 96-Well Calibration Kit is used to maintain your real-time PCR instrument with standard 96-well block. The kit includes region of interest (ROI), background, and dye calibrations, as well as RNaseP verification

  • The 96-Well Calibration Kit is used to maintain your real-time PCR instrument with standard 96-well block.
  • The kit includes region of interest (ROI), background, and dye calibrations, as well as RNaseP verification.
  • Please see your instrument user guide for full instructions on use.
TRUSTED_SUSTAINABILITY

Specifications

Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Reaction Speed Standard
Product Type Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 x 96 well plate
Calibration Type Spectral Calibration
PCR Method qPCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.