missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calmagite, pure, indicator grade

Product Code. 10080730 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10080730 25 g Glass bottle 25g
10458440 5 g Glass bottle 5g
10335980 10 g Glass bottle 10g
10247263 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10080730 Supplier Thermo Scientific Chemicals Supplier No. 108170250
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3147-14-6 | C17H14N2O5S | 358.368 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3147-14-6
Molecular Formula C17H14N2O5S
Molecular Weight (g/mol) 358.368
MDL Number MFCD00011656
InChI Key ASFVMSDYPYMUFL-UHFFFAOYSA-N
Synonym 3-Hydroxy-4-(2-hydroxy-5-methylphenylazo)-1-naphthalenesulfonic acid
PubChem CID 6364506
IUPAC Name 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
SMILES CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Specific Extinction 300 min. (610nm, in buffer)
Suitability suitable for determination of calcium and magnesium
Merck Index 15, 1720
Color Black
Quantity 25 g
Physical Form Crystalline Powder
Formula Weight 358.38
Grade Extra Pure
Chemical Name or Material Calmagite
Show More Show Less
compliance-icons
Product Identifier
  • Calmagite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.