missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calmagite, indicator grade

Product Code. 11427024
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11427024

Brand: Thermo Scientific Chemicals B21482.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3147-14-6 | C17H14N2O5S | 358.368 g/mol

Calmagite is widely used in analytical chemistry as an indicator in the titration of calcium or magnesium with ethylenediaminetetraacetic acid (EDTA). It is also useful in the determination of magnesium in biological materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calmagite is widely used in analytical chemistry as an indicator in the titration of calcium or magnesium with ethylenediaminetetraacetic acid (EDTA). It is also useful in the determination of magnesium in biological materials.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3147-14-6
Molecular Formula C17H14N2O5S
Molecular Weight (g/mol) 358.368
MDL Number MFCD00011656
InChI Key ASFVMSDYPYMUFL-UHFFFAOYSA-N
Synonym 3-Hydroxy-4-(2-hydroxy-5-methylphenylazo)-1-naphthalenesulfonic acid
PubChem CID 6364506
IUPAC Name 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
SMILES CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O

Specifications

Melting Point >330°C
Beilstein 708596
Merck Index 14,1718
Solubility Information Soluble in water.
Formula Weight 358.37
Grade Indicator
Quantity 100 g
Chemical Name or Material Calmagite

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.